(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
Thermo Scientific Chemicals

(S)-(+)-2-Amino-1-propanol, 98%

CAS: 2749-11-3 | C3H9NO | 75.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 104450010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2749-11-3
IUPAC Name(2S)-1-hydroxypropan-2-aminium
Molecular FormulaC3H10NO
InChI KeyBKMMTJMQCTUHRP-VKHMYHEASA-O
SMILESC[C@H]([NH3+])CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration with HClO4>=97.5 %
TLC>=97.5 %
Refractive index1.4480 to 1.4510 (20°C, 589 nm)
Specific optical rotation+20° to +24° (20°C, 589 nm) (c=2, EtOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only