2-Aminopyrimidine, 98%
2-Aminopyrimidine, 98%
2-Aminopyrimidine, 98%
Thermo Scientific Chemicals

2-Aminopyrimidine, 98%

CAS: 109-12-6 | C4H5N3 | 95.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 104581000
Price (USD)
41.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.50
Save 6.85
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.50
Save 6.85
Each
Chemical Identifiers
CAS109-12-6
IUPAC Namepyrimidin-2-amine
Molecular FormulaC4H5N3
InChI KeyLJXQPZWIHJMPQQ-UHFFFAOYSA-N
SMILESNC1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
Melting point123°C to 127°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only