When should I use heat-inactivated serum?
Heat-inactivation of serum inactivates the complements in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.