L(+)-Ascorbic Acid, 99%
L(+)-Ascorbic Acid, 99%
L(+)-Ascorbic Acid, 99%
Thermo Scientific Chemicals

L(+)-Ascorbic Acid, 99%

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 105020025
Price (USD)
343.65
Special offer
Online exclusive
Ends: 28-Dec-2026
404.00
Save 60.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
343.65
Special offer
Online exclusive
Ends: 28-Dec-2026
404.00
Save 60.35
Each
Chemical Identifiers
CAS50-81-7
IUPAC Name(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular FormulaC6H8O6
InChI KeyCIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILESOC[C@H](O)[C@H]1OC(=O)C(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(5 % in water) Passes test
Impurity=<0.2 % Oxalic acid
Iron (Fe)=<2 ppm
Appearance (Form)Crystalline powder or crystals
Titration Iodimetric>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only