Benzethonium chloride, 97%
Benzethonium chloride, 97%
Benzethonium chloride, 97%
Thermo Scientific Chemicals

Benzethonium chloride, 97%

Benzethonium chloride, 97%, C27H42ClNO2, CAS Number-121-54-0, benzethoniumchloride, benzethonium chloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin, 2.5kg, 96% min. (on dry substance) (Argentometry), 100.0, 96.0, CHEBI:31264, White, 204-479-9, 01,433 | CAS: 121-54-0 | C27H42ClNO2 | 448.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 105381000
Price (USD)
70.20
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
70.20
Each
Chemical Identifiers
CAS121-54-0
IUPAC Namebenzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
Molecular FormulaC27H42ClNO2
InChI KeyUREZNYTWGJKWBI-UHFFFAOYSA-M
SMILES[Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Titration Argentometric96.0 to 104.0 % (on dry substance)
Loss on drying=<5 % (105°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only