Benzil, 99+%
Benzil, 99+%
Benzil, 99+%
Thermo Scientific Chemicals

Benzil, 99+%

CAS: 134-81-6 | C14H10O2 | 210.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 105410025
Price (USD)
324.65
Special offer
Online exclusive
Ends: 28-Dec-2026
382.00
Save 57.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
324.65
Special offer
Online exclusive
Ends: 28-Dec-2026
382.00
Save 57.35
Each
Chemical Identifiers
CAS134-81-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point93°C to 97°C
Appearance (Color)Yellow
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only