2,3-Benzofuran, 99%
2,3-Benzofuran, 99%
2,3-Benzofuran, 99%
Thermo Scientific Chemicals

2,3-Benzofuran, 99%

CAS: 271-89-6 | C8H6O | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 105480500
Price (USD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS271-89-6
IUPAC Name1-benzofuran
Molecular FormulaC8H6O
InChI KeyIANQTJSKSUMEQM-UHFFFAOYSA-N
SMILESO1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange-brown
Infrared spectrumConforms
Refractive index1.5655 to 1.5685 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only