Benzonitrile, 99%, pure
Benzonitrile, 99%, pure
Benzonitrile, 99%, pure
Thermo Scientific Chemicals

Benzonitrile, 99%, pure

CAS: 100-47-0 | C7H5N | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 105540025
Price (USD)
136.65
Online exclusive
152.00
Save 15.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
136.65
Online exclusive
152.00
Save 15.35
Each
Chemical Identifiers
CAS100-47-0
IUPAC Namebenzonitrile
Molecular FormulaC7H5N
InChI KeyJFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5270 to 1.5290 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only