Optical Adhesive Cover Plate - FAQs
Optical Adhesive Cover Plate - FAQs
View additional product information for Optical Adhesive Cover Plate - FAQs (10577)
1 product FAQs found
Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?
No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.