2-Benzoylpyridine, 99+%
2-Benzoylpyridine, 99+%
2-Benzoylpyridine, 99+%
Thermo Scientific Chemicals

2-Benzoylpyridine, 99+%

CAS: 91-02-1 | C12H9NO | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 105800250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS91-02-1
IUPAC Name2-benzoylpyridine
Molecular FormulaC12H9NO
InChI KeyGCSHUYKULREZSJ-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point41°C to 45°C
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only