Plasmid pCMV·SPORT-βgal - FAQs
Plasmid pCMV·SPORT-βgal - FAQs
View additional product information for Plasmid pCMV·SPORT-βgal - FAQs (10586014)
4 product FAQs found
How do I resuspend IPTG and X-Gal?
IPTG can be reconstituted in water. Make a stock of 100 mM in water and store working aliquots at -20°C. X-gal can be reconstituted in DMSO, or in a 50:50 mix of DMSO and water. To do the latter, you must dissolve in DMSO first, and then add water to bring up to final volume. It is not necessary to filter sterilize these solutions.
The X-gal solution should be protected from light. To make plates, add 50 ug/ml X-gal and 1 mM (0.24 mg/mL) IPTG to LB/agar that has been cooled down to 50°C. To spread on top of plates, use 50 µl 2% stock of X-gal and 30 µl 100 mM stock of IPTG. 
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?
Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.
Are your vectors routinely sequenced?
No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.
How was the reference sequence for your vectors created?
Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.