Benzyl bromide, 98%
Benzyl bromide, 98%
Benzyl bromide, 98%
Thermo Scientific Chemicals

Benzyl bromide, 98%

CAS: 100-39-0 | C7H7Br | 171.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
Catalog number 105870010
Price (USD)
331.65
Special offer
Online exclusive
Ends: 28-Dec-2026
390.00
Save 58.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
331.65
Special offer
Online exclusive
Ends: 28-Dec-2026
390.00
Save 58.35
Each
Chemical Identifiers
CAS100-39-0
IUPAC Name(bromomethyl)benzene
Molecular FormulaC7H7Br
InChI KeyAGEZXYOZHKGVCM-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5745 to 1.5765 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only