Benzyl 4-hydroxyphenyl ketone, 97%
Benzyl 4-hydroxyphenyl ketone, 97%
Benzyl 4-hydroxyphenyl ketone, 97%
Thermo Scientific Chemicals

Benzyl 4-hydroxyphenyl ketone, 97%

CAS: 2491-32-9 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 105980050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2491-32-9
IUPAC Name1-(4-hydroxyphenyl)-2-phenylethan-1-one
Molecular FormulaC14H12O2
InChI KeyJBQTZLNCDIFCCO-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Appearance (Form)Crystalline powder
Melting point147°C to 151°C
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only