Search
View additional product information for Custom RNA, Tube - FAQs (10620310)
1 product FAQs found
You can order miRNA controls as a custom RNA order at this link. For TaqMan miRNA assays, order the RNA with no modifications. For TaqMan Advanced miRNA assays, order the RNA with a 5' phosphate modification.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center