Benzyltriphenylphosphonium Chloride, 99%
Benzyltriphenylphosphonium Chloride, 99%
Benzyltriphenylphosphonium Chloride, 99%
Thermo Scientific Chemicals

Benzyltriphenylphosphonium Chloride, 99%

CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 106210250
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.20
Save 5.55
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.20
Save 5.55
Each
Chemical Identifiers
CAS1100-88-5
IUPAC Namebenzyltriphenylphosphanium chloride
Molecular FormulaC25H22ClP
InChI KeyUSFRYJRPHFMVBZ-UHFFFAOYSA-M
SMILES[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only