Biphenyl, 99%
Biphenyl, 99%
Biphenyl, 99%
Thermo Scientific Chemicals

Biphenyl, 99%

CAS: 92-52-4 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 106252500
Price (USD)
25.40
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
25.40
Each
Chemical Identifiers
CAS92-52-4
IUPAC Name1,1'-biphenyl
Molecular FormulaC12H10
InChI KeyZUOUZKKEUPVFJK-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals or flakes
Melting point68°C to 72°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only