Search
View additional product information for SuperScript® Rat Brain cDNA Library (8-Week Male, Sprague-Dawley) - FAQs (10653012)
4 product FAQs found
In addition to the CAAT and TATA boxes, the CMV promoter in pcDNA3.1 vector contains sequence homologous to base pairs 137 to 724 of the sequence submitted by Boshart, et al (GenBank Accession # K03104). The complete enhancer region is contained between 214 and 620 of this sequence. Therefore, by this definition, the CMV promoter could be said to be "complete".
Please note that this promoter does not contain an intron. Some people believe that the complete promoter must contain the intron, but that has not been demonstrated to be necessary for expression.
It depends upon the library. Only libraries in pCMVSport6 or pCMVSport6.1 have attB sequences.
Yes, but the pCMVSport vector does not have a eukaryotic origin of replication or selectable marker, so you can only screen for transient expression.
You can only do this if you transfect the DNA into eukaryotic cells. There is no bacterial promoter in the pCMVSport vector.