2-Bromoethanol, 97%
2-Bromoethanol, 97%
2-Bromoethanol, 97%
Thermo Scientific Chemicals

2-Bromoethanol, 97%

CAS: 540-51-2 | C2H5BrO | 124.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
250 mL
Packaging:
Glass bottle
Catalog number 106920050
Price (USD)
71.65
Online exclusive
79.60
Save 7.95
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Online exclusive
79.60
Save 7.95
Each
Chemical Identifiers
CAS540-51-2
IUPAC Name2-bromoethan-1-ol
Molecular FormulaC2H5BrO
InChI KeyLDLCZOVUSADOIV-UHFFFAOYSA-N
SMILESOCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light orange
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4890 to 1.4940 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only