Custom DNA Oligos, Order Tubes - FAQs
Custom DNA Oligos, Order Tubes - FAQs
View additional product information for Custom DNA Oligos, Order Tubes - FAQs (10693018)
1 product FAQs found
What scales of synthesis do you offer for custom DNA oligos?
For custom DNA oligos, we offer 25 nmol, 50 nmol, 200 nmol, 1 µmol, or 10 µmol. These amounts are the starting point for synthesis, not the guaranteed final amount. Oligonucleotide length, sequence, GC content, and coupling efficiencies can vary the yield of nmole amounts post‐synthesis.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center