(2-Bromoethyl)benzene, 98%
(2-Bromoethyl)benzene, 98%
(2-Bromoethyl)benzene, 98%
Thermo Scientific Chemicals

(2-Bromoethyl)benzene, 98%

CAS: 103-63-9 | C8H9Br | 185.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 106945000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-63-9
IUPAC Name(2-bromoethyl)benzene
Molecular FormulaC8H9Br
InChI KeyWMPPDTMATNBGJN-UHFFFAOYSA-N
SMILESBrCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only