N-(2-Bromoethyl)phthalimide, 97%
N-(2-Bromoethyl)phthalimide, 97%
N-(2-Bromoethyl)phthalimide, 97%
Thermo Scientific Chemicals

N-(2-Bromoethyl)phthalimide, 97%

CAS: 574-98-1 | C10H8BrNO2 | 254.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 106971000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS574-98-1
IUPAC Name2-(2-bromoethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC10H8BrNO2
InChI KeyCHZXTOCAICMPQR-UHFFFAOYSA-N
SMILESBrCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 % (on dry substance)
Water=<1 % (K.F.)
Infrared spectrumConforms
Appearance (Color)White to beige to light pink
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only