1-Bromo-2-naphthol, 98%
1-Bromo-2-naphthol, 98%
1-Bromo-2-naphthol, 98%
Acros Organics

1-Bromo-2-naphthol, 98%

CAS: 573-97-7 | C10H7BrO | 223.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 107160250
also known as 10716
HSN 29081900
Price (INR)
9,436.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,436.00
Each
Chemical Identifiers
CAS573-97-7
IUPAC Name1-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyFQJZPYXGPYJJIH-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Off-white to beige to brown-purple
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point77°C to 84°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only