Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride - FAQs
Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride - FAQs
View additional product information for Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride - FAQs (10725018)
2 product FAQs found
Which serum-free media can be used to culture mouse keratinocytes?
The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
For how long can I store Keratinocyte-SFM at 4 degrees C?
The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.