DL-2-Bromopropionic acid, 99%
DL-2-Bromopropionic acid, 99%
DL-2-Bromopropionic acid, 99%
Thermo Scientific Chemicals

DL-2-Bromopropionic acid, 99%

CAS: 598-72-1 | C3H5BrO2 | 152.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 107350010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS598-72-1
IUPAC Name2-bromopropanoic acid
Molecular FormulaC3H5BrO2
InChI KeyMONMFXREYOKQTI-UHFFFAOYNA-N
SMILESCC(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
HPLC>=98.5 %
Appearance (Form)Liquid after melting
Refractive index1.4740 to 1.4760 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only