3-Bromopropylamine hydrobromide, 98%
3-Bromopropylamine hydrobromide, 98%
3-Bromopropylamine hydrobromide, 98%
Thermo Scientific Chemicals

3-Bromopropylamine hydrobromide, 98%

CAS: 5003-71-4 | C3H8BrN·HBr | 218.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
10738025025 gGlass bottle
Catalog number 107380250
Price (USD)
53.40
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.40
Each
Chemical Identifiers
CAS5003-71-4
IUPAC Namehydrogen 3-bromopropan-1-amine bromide
Molecular FormulaC3H9Br2N
InChI KeyPQIYSSSTRHVOBW-UHFFFAOYSA-N
SMILES[H+].[Br-].NCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige to light pink
Appearance (Color)to orange to brown
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only