n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
Thermo Scientific Chemicals

n-Butyl acetate, 99+%, extra pure

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Metal drum
Catalog number 107750250
Price (USD)
798.65
Special offer
Online exclusive
Ends: 28-Dec-2026
939.00
Save 140.35
Each
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (USD)
798.65
Special offer
Online exclusive
Ends: 28-Dec-2026
939.00
Save 140.35
Each
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Water=<0.1 % (K.F.)
Appearance (Color)Clear colorless
Refractive index1.3930 to 1.3950 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only