tert-Butyl acetoacetate, 97%
tert-Butyl acetoacetate, 97%
tert-Butyl acetoacetate, 97%
Thermo Scientific Chemicals

tert-Butyl acetoacetate, 97%

CAS: 1694-31-1 | C8H14O3 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 107785000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1694-31-1
IUPAC Nametert-butyl 3-oxobutanoate
Molecular FormulaC8H14O3
InChI KeyJKUYRAMKJLMYLO-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4180 to 1.4210 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only