tert-Butylbenzene, 99%
tert-Butylbenzene, 99%
tert-Butylbenzene, 99%
Thermo Scientific Chemicals

tert-Butylbenzene, 99%

CAS: 98-06-6 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 107870010
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Chemical Identifiers
CAS98-06-6
IUPAC Nametert-butylbenzene
Molecular FormulaC10H14
InChI KeyYTZKOQUCBOVLHL-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only