n-Butyltriphenylphosphonium bromide, 99%
n-Butyltriphenylphosphonium bromide, 99%
n-Butyltriphenylphosphonium bromide, 99%
Thermo Scientific Chemicals

n-Butyltriphenylphosphonium bromide, 99%

CAS: 1779-51-7 | C22H24BrP | 399.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 108061000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1779-51-7
IUPAC Namebutyltriphenylphosphanium bromide
Molecular FormulaC22H24BrP
InChI KeyIKWKJIWDLVYZIY-UHFFFAOYSA-M
SMILES[Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric>=98.5 %
Appearance (Color)White to off-white
Melting point238°C to 243°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only