Caffeine, 98.5%, specified according to the req. of USP/BP
Caffeine, 98.5%, specified according to the req. of USP/BP
Caffeine, 98.5%, specified according to the req. of USP/BP
Thermo Scientific Chemicals

Caffeine, 98.5%, specified according to the req. of USP/BP

The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC. | CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Quantity:
10 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 108165000
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS58-08-2
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC8H10N4O2
InChI KeyRYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)N(C)C(=O)N2C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or almost white
Loss on drying=<0.5 %
Sulfated ash=<0.1 %
Appearance of solution(0.5g in 50 ml water) Clear colorless
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
RUO – Research Use Only