N-Carbethoxyphthalimide, 99+%
N-Carbethoxyphthalimide, 99+%
N-Carbethoxyphthalimide, 99+%
Thermo Scientific Chemicals

N-Carbethoxyphthalimide, 99+%

CAS: 22509-74-6 | C11H9NO4 | 219.2 g/mol
Have Questions?
Catalog NumberQuantityPackaging
10831050050 gGlass bottle
Catalog number 108310500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS22509-74-6
IUPAC Nameethyl 1,3-dioxo-2,3-dihydro-1H-isoindole-2-carboxylate
Molecular FormulaC11H9NO4
InChI KeyVRHAQNTWKSVEEC-UHFFFAOYSA-N
SMILESCCOC(=O)N1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point82°C to 84°C
Water=<0.5 % (K.F.)
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only