Methyl succinyl chloride, 97%
Methyl succinyl chloride, 97%
Methyl succinyl chloride, 97%
Thermo Scientific Chemicals

Methyl succinyl chloride, 97%

CAS: 1490-25-1 | C5H7ClO3 | 150.56 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 108400050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1490-25-1
IUPAC Namemethyl 4-chloro-4-oxobutanoate
Molecular FormulaC5H7ClO3
InChI KeySRXOJMOGPYFZKC-UHFFFAOYSA-N
SMILESCOC(=O)CCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only