Carboxymethoxylamine HemiHydroChloride, 98%
Carboxymethoxylamine HemiHydroChloride, 98%
Carboxymethoxylamine HemiHydroChloride, 98%
Thermo Scientific Chemicals

Carboxymethoxylamine HemiHydroChloride, 98%

CAS: 2921-14-4 | C4H11ClN2O6 | 218.59 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
產品號碼 108430025
價格 (TWD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2921-14-4
規格Specification Sheet規格表
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only