DL-Carnitine hydrochloride, 99%
DL-Carnitine hydrochloride, 99%
DL-Carnitine hydrochloride, 99%
Thermo Scientific Chemicals

DL-Carnitine hydrochloride, 99%

CAS: 461-05-2 | C7H15NO3·HCl | 197.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 108450250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS461-05-2
IUPAC Name(3-carboxy-2-hydroxypropyl)trimethylazanium chloride
Molecular FormulaC7H16ClNO3
InChI KeyJXXCENBLGFBQJM-UHFFFAOYNA-N
SMILES[Cl-].C[N+](C)(C)CC(O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point194°C to 200°C
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with HClO4>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only