Product Image
Thermo Scientific™

DRI™ Acetaminophen Serum Toxicology Assay

Identify the overdose class of drug in emergency room situations where urine samples are not readily available with this Acetaminophen serum toxicology assay.
Have Questions?
Catalog NumberQuantity
108625 x 8 mL
Catalog number 1086
Price (USD)
-
Quantity:
25 x 8 mL

Identify the overdose class of drug in emergency room situations where urine samples are not readily available with Thermo Scientific™ DRI™ Acetaminophen Serum Toxicology Assay. Provided in convenient packaging with separate calibrators, these liquid, ready-to-use assays have the flexibility to run on a variety of clinical chemistry analyzers, providing objective results.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionAcetaminophen Assay
Quantity25 x 8 mL
Sample TypeSerum
TypeReagent
Detectable AnalytesAcetaminophen
Unit SizeEach
Contents & Storage
2°C to 8°C