2-Chlorobenzonitrile, 99%
2-Chlorobenzonitrile, 99%
2-Chlorobenzonitrile, 99%
Thermo Scientific Chemicals

2-Chlorobenzonitrile, 99%

CAS: 873-32-5 | C7H4ClN | 137.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 108672500
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.80
Save 11.15
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.80
Save 11.15
Each
Chemical Identifiers
CAS873-32-5
IUPAC Name2-chlorobenzonitrile
Molecular FormulaC7H4ClN
InChI KeyNHWQMJMIYICNBP-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)chunks or solid
Melting point43°C to 46°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only