4-Chlorocinnamic acid, 99%, predominantly trans
4-Chlorocinnamic acid, 99%, predominantly trans
4-Chlorocinnamic acid, 99%, predominantly trans
Thermo Scientific Chemicals

4-Chlorocinnamic acid, 99%, predominantly trans

CAS: 1615-02-7 | C9H7ClO2 | 182.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 108940250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1615-02-7
SpecificationsSpecification SheetSpecification Sheet
Melting point246°C to 250°C
Titration with NaOH98.5 to 101.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only