1-Chloro-3-hydroxypropane, 98%, stabilized
1-Chloro-3-hydroxypropane, 98%, stabilized
1-Chloro-3-hydroxypropane, 98%, stabilized
Thermo Scientific Chemicals

1-Chloro-3-hydroxypropane, 98%, stabilized

CAS: 627-30-5 | C3H7ClO | 94.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 109290250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS627-30-5
IUPAC Name3-chloropropan-1-ol
Molecular FormulaC3H7ClO
InChI KeyLAMUXTNQCICZQX-UHFFFAOYSA-N
SMILESOCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
Stabilizer0.1 % Na2CO3
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only