5-Chloro-8-hydroxyquinoline, 98%
5-Chloro-8-hydroxyquinoline, 98%
5-Chloro-8-hydroxyquinoline, 98%
Thermo Scientific Chemicals

5-Chloro-8-hydroxyquinoline, 98%

CAS: 130-16-5 | C9H6ClNO | 179.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass bottle
Catalog number 109300050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS130-16-5
IUPAC Name5-chloroquinolin-8-ol
Molecular FormulaC9H6ClNO
InChI KeyCTQMJYWDVABFRZ-UHFFFAOYSA-N
SMILESOC1=C2N=CC=CC2=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)to light brown
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to beige to light green to grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only