5-Chloroindole, 99%
5-Chloroindole, 99%
5-Chloroindole, 99%
Thermo Scientific Chemicals

5-Chloroindole, 99%

CAS: 17422-32-1 | C8H6ClN | 151.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 109310050
Price (EUR)
176,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
176,00
Each
Chemical Identifiers
CAS17422-32-1
IUPAC Name5-chloro-1H-indole
Molecular FormulaC8H6ClN
InChI KeyMYTGFBZJLDLWQG-UHFFFAOYSA-N
SMILESClC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
GC>=98.5 %
Appearance (Form)Crystalline powder
Melting point69°C to 72°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only