1-Chloro-4-nitrobenzene, 99%
1-Chloro-4-nitrobenzene, 99%
1-Chloro-4-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Chloro-4-nitrobenzene, 99%

CAS: 100-00-5 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number 109630050
Price (EUR)
24,65
Online exclusive
26,70
Save 2,05 (8%)
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
24,65
Online exclusive
26,70
Save 2,05 (8%)
Each
Specifications
Assay Percent Range98.5% min. (GC)
Beilstein05, 243
CAS Max %100.0
CAS Min %98.5
CAS100-00-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only