Product Image
Gibco™

Sf-900™ Medium (1.3X)

Sf-900™ SFM is a complete serum-free, low protein insect cell culture medium developed for the convenient and reproducible formation ofRead more
Have Questions?
Catalog NumberQuantity
10967032100 mL
Catalog number 10967032
Price (MXN)
-
Quantity:
100 mL
Sf-900™ SFM is a complete serum-free, low protein insect cell culture medium developed for the convenient and reproducible formation of 1% agarose overlays used for plaque assays of baculovirus in Spodoptera frugiperda (Sf9 and Sf21) cells. When used with Gibco™ 4% Agarose Gel, Sf900™ SFM (1.3x) allows for convenient formation of a 1% overlay with correct 1x final osmolality and nutrition required for insect cell growth during the plaque assay.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco™ Sf-900 II SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineSf21, Sf9
Cell TypeInsect Cell
Product LineGibco, Sf-900
Product TypeInsect Cell Serum Free Medium (SFM)
Quantity100 mL
Shipping ConditionRoom Temperature
SpeciesS. frugiperda, Spodoptera frugiperda
FormLiquid
With AdditivesGlutamine
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 9 months from date of manufacture

Frequently asked questions (FAQs)

What medium do you recommend to grow Sf9 or Sf21 cells for performing plaque assays?

We would recommend using our Grace's Supplemented Insect Medium (Cat. No. 11667037) or SF-900 Medium (1.3X) (Cat. No. 10967032). Grace's Supplmented Insect Media is a 2X formulation intended for use in the agarose overlay when performing the plaque assay. The media has been modified with:
- Lactalbumin hydrolysate
- Yeastolate
- L-glutamine
- Sodium bicarbonate

Please go here for the complete formulation (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.72.html). SF-900 Medium (1.3X) is a complete serum-free low protein insect cell culture medium for Sf9 and Sf21 cells, which can be used for 1% agarose overlays used for plaque assays.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.