2-Chloro-3-nitropyridine, 99+%
2-Chloro-3-nitropyridine, 99+%
2-Chloro-3-nitropyridine, 99+%
Thermo Scientific Chemicals

2-Chloro-3-nitropyridine, 99+%

CAS: 5470-18-8 | C5H3ClN2O2 | 158.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 109730250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5470-18-8
IUPAC Name2-chloro-3-nitropyridine
Molecular FormulaC5H3ClN2O2
InChI KeyUUOLETYDNTVQDY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Melting point100°C to 103°C
Infrared spectrumConforms
HPLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only