UltraPure™ DNase/RNase-Free Distilled Water
Invitrogen™
UltraPure™ DNase/RNase-Free Distilled Water
UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 x 500 mL
Catalog number 10977049
Price (USD)
***
Quantity:
10 x 500 mL
Request bulk or custom format
Price (USD)
***
UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.
Performance and quality testing
No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineUltraPure
Purification MethodMembrane-Filtered
Shipping ConditionRoom Temperature
Treatment(s)Not DEPC-Treated
GradeMolecular Biology
Quantity10 x 500 mL
Unit SizeEach