alpha-Chloro-o-xylene, 99%
alpha-Chloro-o-xylene, 99%
alpha-Chloro-o-xylene, 99%
Acros Organics

alpha-Chloro-o-xylene, 99%

CAS: 552-45-4 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 110150250
also known as 11015
HSN 29039990
Price (INR)
5,065.00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,065.00
Each
Chemical Identifiers
CAS552-45-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.5405 to 1.5425 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only