Cholesteryl benzoate, 98%
Cholesteryl benzoate, 98%
Cholesteryl benzoate, 98%
Thermo Scientific Chemicals

Cholesteryl benzoate, 98%

CAS: 604-32-0 | C34H50O2 | 490.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 110200250
Price (USD)
50.65
Special offer
Online exclusive
Ends: 28-Dec-2026
59.80
Save 9.15
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
50.65
Special offer
Online exclusive
Ends: 28-Dec-2026
59.80
Save 9.15
Each
Chemical Identifiers
CAS604-32-0
IUPAC Name9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl benzoate
Molecular FormulaC34H50O2
InChI KeyUVZUFUGNHDDLRQ-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only