AlbuMax I Lipid Rich BSA
Gibco™

AlbuMax I Lipid Rich BSA

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤0.1%. Lyophilized powder.
Have Questions?
Catalog NumberQuantity
11020062100 g
Catalog number 11020062
Price (USD)
-
Quantity:
100 g

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤0.1%. Lyophilized powder.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications
Cell TypeMammalian Cells
ColorBeige
Product TypeBSA
Quantity100 g
SterilityNon-sterile
Storage Requirements2°C to 8°C
FormSolid
Unit SizeEach

Frequently asked questions (FAQs)

What fatty acids are in my specific lot of AlbuMAX I? What are their approximate concentrations?

Below are the concentrations of free fatty acids obtained from the HPLC lipid profiles of an older lot of AlbuMAX I:

Alpha-linolenic acid: 0.59 mg FFA/g protein
Linoleic acid: 0.65 mg FFA/g protein
Oleic acid: 2.12 mg FFA/g protein
Stearic acid: 2.09 mg FFA/g protein
Palmitic acid: 1.90 mg FFA/g protein
Note: The individual lipid concentrations may vary between lots, but the values above can be used as a good approximation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.