AlbuMax II Lipid Rich BSA
Actual product may vary
AlbuMax II Lipid Rich BSA
AlbuMax II Lipid Rich BSA
Gibco™

AlbuMax II Lipid Rich BSA

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. Lyophilized powder.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
110210451 kg
110210115 g
110210865 kg
Catalog number 11021045
Price (USD)
3,680.00
1 kg
Quantity:
1 kg
Price (USD)
3,680.00
1 kg

This is a lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤ 1%.

Origin: New Zealand

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications
Product LineAlbuMAX, Gibco
Product TypeBSA
Quantity1 kg
Shipping ConditionRoom Temperature
Unit Size1 kg
Contents & Storage
Store in refrigerator (2°C to 8°C).