1,8-Cineole, 99%
1,8-Cineole, 99%
1,8-Cineole, 99%
Thermo Scientific Chemicals

1,8-Cineole, 99%

CAS: 470-82-6 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 110340050
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.70
Save 4.05
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.70
Save 4.05
Each
Chemical Identifiers
CAS470-82-6
IUPAC Name1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
Molecular FormulaC10H18O
InChI KeyWEEGYLXZBRQIMU-UHFFFAOYSA-N
SMILESCC12CCC(CC1)C(C)(C)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Solubility(in ethanol (70% v/v)) 1:2.5
Appearance (Color)Clear colorless to light yellow
Refractive index1.4550 to 1.4700 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only