trans-Cinnamaldehyde, 99%
trans-Cinnamaldehyde, 99%
trans-Cinnamaldehyde, 99%
Thermo Scientific Chemicals

trans-Cinnamaldehyde, 99%

CAS: 14371-10-9 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 110355000
Price (USD)
56.10
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
56.10
Each
Chemical Identifiers
CAS14371-10-9
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6210 to 1.6230 (20°C, 589 nm)
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only